missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C9orf64 (aa 82-166) Control Fragment Recombinant Protein

Catalog No. RP93035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93035 Supplier Invitrogen™ Supplier No. RP93035

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63038. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Involved in salvaging queuosine.

Specifications

Accession Number Q5T6V5
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 84267
Name Human C9orf64 (aa 82-166) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C9orf64; chromosome 9 open reading frame 64; Queuosine salvage protein; UPF0553 protein; UPF0553 protein C9orf64
Common Name C9orf64
Gene Symbol C9orf64
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QDEHKCVVRYRGKTYSGYWSLCAAVNRALDEGIPITSASYYATVTLDQVRNILRSDTDVSMPLVEERHRILNETGKILLEKFGGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less