missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C6orf129 (aa 4-42) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100013
Description
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60953 (PA5-60953. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CCDC167 is a protein coding gene.Specifications
| Q9P0B6 | |
| Blocking Assay, Control | |
| 154467 | |
| 100 μL | |
| 1110021J02Rik; C23H6orf129; C6orf129; CCDC167; coiled-coil domain containing 167; coiled-coil domain-containing protein 167; HSPC265; transmembrane and coiled-coil domain-containing protein C6orf129; transmembrane and coiled-coil domain-containing protein C6orf129 homolog | |
| CCDC167 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human C6orf129 (aa 4-42) Control Fragment | |
| RUO | |
| C6orf129 | |
| Unconjugated | |
| Recombinant | |
| KKRENLGVALEIDGLEEKLSQCRRDLEAVNSRLHSRELS | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |