missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human C5a, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF30070500
Encompass
Change view
Click to view available options
Quantity:
100 μg
20 μg
500 μg
1 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF30070500 500 μg
AF30070100 100 μg
AF300701MG 1 mg
AF3007020UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AF30070500 Supplier Gibco™ Supplier No. AF30070500UG

Recombinant Protein

Recombinant Human C5a is an 8.3 kDa glycoprotein containing the 74 amino acid residues of the C5a anaphylatoxin chain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is the fifth component of complement, which plays an important role in inflammatory and cell killing processes. This protein is comprised of alpha and beta polypeptide chains that are linked by a disulfide bridge. An activation peptide, C5a, which is an anaphylatoxin that possesses potent spasmogenic and chemotactic activity, is derived from the alpha polypeptide via cleavage with a convertase. The C5b macromolecular cleavage product can form a complex with the C6 complement component, and this complex is the basis for formation of the membrane attack complex, which includes additional complement components. Mutations in this gene cause complement component 5 deficiency, a disease where patients show a propensity for severe recurrent infections. Defects in this gene have also been linked to a susceptibility to liver fibrosis and to rheumatoid arthritis.

Specifications

Accession Number P01031
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 727
Molecular Weight (g/mol) 8.3 kDa
Name Human C5a, Animal-Free
Quantity 500 μg
Source E. coli
Endotoxin Concentration <0.1 EU/ μg
Gene Alias anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905
Common Name Complement C5a
Gene Symbol C5
Biological Activity Measured by its ability to support adhesion of Human Umbilical Vein Endothelial Cells (HUVEC) to the Recombinant Human C5a-coated surface.
Conjugate Unconjugated
Recombinant Recombinant
Sequence TLQKKIEEIA AKYKHSVVKK CCYDGACVNN DETCEQRAAR ISLGPRCIKA FTECCVVASQ LRANISHKDM QLGR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less