missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Complement C3a ELISA Kit

Catalog No. 501125296
Change view
Click to view available options
Quantity:
2 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125296 2 x 96 Tests
501125293 96 Tests
501125294 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 501125296 Supplier Invitrogen™ Supplier No. BMS2089TWO
Only null left

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human C3a

ELISA

The Human Complement C3a (Hu C3a) ELISA quantitates Hu C3a in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu C3a. Principle of the method The Human C3a solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.

Specifications

Accession Number P01024
Assay Range 0.3 to 20 ng/mL
Assay Sensitivity 0.14 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 718
Gene Symbol AHUS5,ARMD9,ASPa,C3b,CPAMD1,HEL-S-62 p,C3
Interassay CV 5.9%
Intraassay CV 5.9
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 10 μL, Serum 10 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Synonym complement component 3, ASP, C3a, C3b, C3, Acylation stimulating protein
Target Complement C3a
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 30 mins
Show More Show Less