missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C3 native Protein from human plasma
Used for SDS-PAGE
Supplier: Abnova™ P4945
Description
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection. [provided by RefSeq]
Specifications
| SDS-PAGE | |
| 718 | |
| Native protein purified from human plasma | |
| Store at -80°C. After reconstitution with 100 uL deionized water, store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| ARMD9/ASP/CPAMD1 | |
| C3 | |
| None | |
| Lyophilized |
| Lyophilized from Sodium phosphate buffer, pH 7.2 (0.15M NaCl). | |
| C3 (Human) Native Protein | |
| 100 μg | |
| RUO | |
| C3 | |
| Human | |
| Native protein purified from human plasma | |
| >90% by SDS-PAGE |