missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C21orf91 (aa 183-294) Control Fragment Recombinant Protein

Catalog No. RP91810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91810 Supplier Invitrogen™ Supplier No. RP91810

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53773 (PA5-53773. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

C21orf91 is a protein coding gene.

Specifications

Accession Number Q9NYK6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54149
Name Human C21orf91 (aa 183-294) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C21orf14; C21orf38; C21orf91; chromosome 21 open reading frame 91; cold sore susceptibility gene 1; CSSG1; early undifferentiated retina and lens; EURL; Protein EURL homolog; YG81
Common Name C21orf91
Gene Symbol C21orf91
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CRNSVLWPHSHNQAQKKEETISSPEANVQTQHPHYSREELNSMTLGEVEQLNAKLLQQIQEVFEELTHQVQEKDSLASQLHVRHVAIEQLLKNCSKLPCLQVGRTGMKSHLP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less