missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C19orf66 Control Fragment Recombinant Protein

Catalog No. RP99101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99101 Supplier Invitrogen™ Supplier No. RP99101

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59815 (PA5-59815. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Inhibits programmed -1 ribosomal frameshifting (-1PRF) of a variety of mRNAs from viruses, such as HIV1, and cellular genes, such as PEG10. Interacts with the -1PRF signal of target mRNA and translating ribosomes and causes premature translation termination at the frameshifting site (PubMed:30682371). Regulates HIV1 GAG-POL expression by inhibiting -1PRF (PubMed:30682371). Exhibits antiviral activity against dengue virus (DENV) and can inhibit the replication of all DENV serotypes. May block the protein translation of DENV RNA via its association with cellular mRNA-binding proteins and viral RNA. Can also limit the replication of hepatitis C virus (HCV), West Nile virus (WNV), Chikungunya virus (CHIKV), herpes simplex virus type 1 (HHV-1) and human adenovirus (PubMed:26735137, PubMed:27974568). Binds nucleic acids with a higher affinity for ssRNA and ssDNA than for dsDNA (PubMed:27974568). [UniProt]

Specifications

Accession Number Q9NUL5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55337
Name Human C19orf66 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C19orf66; chromosome 19 open reading frame 66; FLJ11286; Interferon-regulated antiviral protein; IRAV; Repressor of yield of Dengue virus; repressor of yield of DENV; repressor of yield of DENV protein; RYDEN; SFL; SHFL; Shiftless antiviral inhibitor of ribosomal frameshifting protein; UPF0515 protein C19orf66
Common Name C19orf66
Gene Symbol SHFL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KFHGKASSKKAGALMRKFGSDHTGVGRSIVYGVKQKDGQELSNDLDAQDPPEDMKQDRDIQAVATSLLPLTEANLRMFQRAQDD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less