missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C17orf89 (aa 46-70) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100584
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63474 (PA5-63474. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
NDUFAF8 is a protein coding gene involved in the assembly of mitochondrial NADH:ubiquinone oxidoreductase complex (complex I, MT-ND1). Required to stabilize NDUFAF5.Specifications
| A1L188 | |
| Blocking Assay, Control | |
| 284184 | |
| 100 μL | |
| 1810043H04Rik; C17orf89; C9H17orf89; chromosome 17 open reading frame 89; hypothetical protein LOC690871; LOC690871; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 8; NADH:ubiquinone oxidoreductase complex assembly factor 8; Ndufaf8; RIKEN cDNA 1810043H04 gene; uncharacterized protein C17orf89; uncharacterized protein C17orf89 homolog | |
| NDUFAF8 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human C17orf89 (aa 46-70) Control Fragment | |
| RUO | |
| C17orf89 | |
| Unconjugated | |
| Recombinant | |
| LSKDFCAREFEALRSCFAAAAKKTL | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |