missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C17orf59 (aa 249-329) Control Fragment Recombinant Protein

Catalog No. RP99998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99998 Supplier Invitrogen™ Supplier No. RP99998

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60993. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

As part of the BORC complex may play a role in lysosomes movement and localization at the cell periphery. Associated with the cytosolic face of lysosomes, the BORC complex may recruit ARL8B and couple lysosomes to microtubule plus-end-directed kinesin motor.

Specifications

Accession Number Q96GS4
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 54785
Name Human C17orf59 (aa 249-329) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2310047M10Rik; BLOC-1 related complex subunit 6; BLOC-1-related complex subunit 6; BORCS6; C17orf59; LOC497934; Lysosome-dispersing protein; Lyspersin; PRO2472; RIKEN cDNA 2310047M10 gene; similar to hypothetical protein FLJ20014; uncharacterized protein C17orf59; uncharacterized protein C17orf59 homolog
Common Name C17orf59
Gene Symbol BORCS6
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PTPAIPPIDPEVLRDLERLSRELGGRVDRLLRGLGGAVQELTALSVGCIQTYRDAVDSLGEAVDMSIKGMYTLLARCEELE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less