missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C17orf51 (aa 75-143) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100564
Description
Highest antigen sequence indentity to the following orthologs: Mouse (25%), Rat (25%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62552 (PA5-62552. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
C17orf51 is a protein coding gene.Specifications
| A8MQB3 | |
| Blocking Assay, Control | |
| 339263 | |
| 100 μL | |
| C17orf51; chromosome 17 open reading frame 51; uncharacterized protein C17orf51 | |
| LINC02693 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human C17orf51 (aa 75-143) Control Fragment | |
| RUO | |
| C17orf51 | |
| Unconjugated | |
| Recombinant | |
| ALYAPALRLRDHLDRFSILMTSCTSWLQAPQAPGLCRDEQSSRISVPQLSGAPILLPDLEGTKLSNFQE | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |