missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C16orf72 (aa 58-148) Control Fragment Recombinant Protein

Catalog No. RP98783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98783 Supplier Invitrogen™ Supplier No. RP98783

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59626 (PA5-59626. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

C16orf72 is a protein coding gene.

Specifications

Accession Number Q14CZ0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29035
Name Human C16orf72 (aa 58-148) Control Fragment
Quantity 100 μL
Gene Alias C16orf72; chromosome 16 open reading frame 72; PRO0149; UPF0472 protein C16orf72
Common Name C16orf72
Gene Symbol C16orf72
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KLWHLFQNSATAVAQLYKDRVCQQPGLSLWVPFQNAATAVTNLYKESVDTHQRSFDIGIQIGYQRRNKDVLAWVKKRRRTIRREDLISFLC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less