missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C15orf40 (aa 28-86) Control Fragment Recombinant Protein

Catalog No. RP110190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP110190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP110190 Supplier Invitrogen™ Supplier No. RP110190

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144798. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

C15orf40 is a protein coding gene.

Specifications

Accession Number Q8WUR7
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 123207
Name Human C15orf40 (aa 28-86) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 3110040N11Rik; AI851475; C15orf40; chromosome 15 open reading frame 40; RIKEN cDNA 3110040N11 gene; UPF0235 protein C15orf40; UPF0235 protein C15orf40 homolog
Common Name C15orf40
Gene Symbol C15orf40
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPKKAGATTKGKSQSKEPERPLPPLGPVAVDPKGCVTIAIHAKPGSKQNAVTDLTAEAV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less