missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C14orf159 (aa 148-248) Control Fragment Recombinant Protein

Catalog No. RP98444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98444 Supplier Invitrogen™ Supplier No. RP98444

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59566 (PA5-59566. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

C14orf159 is a protein coding gene.

Specifications

Accession Number Q7Z3D6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80017
Name Human C14orf159 (aa 148-248) Control Fragment
Quantity 100 μL
Gene Alias C14orf159; chromosome 14 open reading frame 159; DGLUCY; D-glutamate cyclase, mitochondrial; UNQ2439/PRO5000; UPF0317 protein C14orf159, mitochondrial
Common Name C14orf159
Gene Symbol DGLUCY
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AGAYKTTVPCVTHAGFCCPLVVTMRPIPKDKLEGLVRACCSLGGEQGQPVHMGDPELLGIKELSKPAYGDAMVCPPGEVPVFWPSPLTSLGAVSSCETPLA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less