missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C14orf145 (aa 724-863) Control Fragment Recombinant Protein

Catalog No. RP88851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP88851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP88851 Supplier Invitrogen™ Supplier No. RP88851

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53819 (PA5-53819. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CEP128 gene ontology annotations related to this gene include protein localization.

Specifications

Accession Number Q6ZU80
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 145508
Name Human C14orf145 (aa 724-863) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4930534B04Rik; 5430424K18Rik; AI449708; AI480494; C14orf145; C14orf61; centrosomal protein 128; centrosomal protein 128 kDa; centrosomal protein of 128 kDa; Cep128; E230024F20; LEDP/132; lens epithelium-derived protein; LOC500700; similar to chromosome 14 open reading frame 145
Common Name C14orf145
Gene Symbol CEP128
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KEKSEAENHIRTLKAESLEEKNMAKIHRGQLEKLKSQCDRLTEELTQNENENKKLKLKYQCLKDQLEEREKHISIEEEHLRRMEEARLQLKDQLLCLETEQESILGVIGKEIDAACKTFSKDSVEKLKVFSSGPDIHYDP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less