missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C11orf1 (aa 75-150) Control Fragment Recombinant Protein

Catalog No. RP97357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97357 Supplier Invitrogen™ Supplier No. RP97357

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58383 (PA5-58383. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

C11orf1 is a protein coding gene.

Specifications

Accession Number Q9H5F2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64776
Name Human C11orf1 (aa 75-150) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C11orf1; chromosome 11 open reading frame 1; UPF0686 protein; UPF0686 protein C11orf1
Common Name C11orf1
Gene Symbol C11orf1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QERYDLRNIVQPKPLPSQFGHYFETTYDTSYNNKMPLSTHRFKREPHWFPGHQPELDPPRYKCTEKSTYMNSYSKP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less