missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human C1 Inhibitor Recombinant Protein, PeproTech®

Catalog No. 13020500ug
Encompass
Change view
Click to view available options
Quantity:
200 μg
50 μg
500 μg
1 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13020500UG 500 μg
130201MG 1 mg
13020200UG 200 μg
1302050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13020500UG Supplier Gibco™ Supplier No. 13020500UG
Only null left

Recombinant Protein

Recombinant Human C1 Inhibitor is a highly glycosylated glycoprotein containing 478 amino acid residues (52.8kDa), corresponding to amino acids 23-500 of the C1 inhibitor precursor, and is fully functional in its ability to inhibit the C1 complex. Glycosylated C1 Inhibitor migrates at an apparent molecular weight of approximately 80-90 kDa by SDS PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ∽104 kDa. Serum concentration has been determined to 180 μg/mL.

Specifications

Accession Number P05155
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 710
Molecular Weight (g/mol) 52.8 kDa
Name Human C1 Inhibitor
Quantity 500 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1Inh; C1-inhibiting factor; C1nh; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH
Common Name C1 inhibitor
Gene Symbol SERPING1
Biological Activity Measured by its ability to inhibit recombinant human complement component C1a cleavage of a colorimetric peptide substrate, N Carbobenzyloxy-Lys-ThioBenzyl ester (Z-K-SBzl). The expected IC50 is ≤ 2.6 nM.
Conjugate Unconjugated
Recombinant Recombinant
Sequence NPNATSSSSQ DPESLQDRGE GKVATTVISK MLFVEPILEV SSLPTTNSTT NSATKITANT TDEPTTQPTT EPTTQPTIQP TQPTTQLPTD SPTQPTTGSF CPGPVTLCSD LESHSTEAVL GDALVDFSLK LYHAFSAMKK VETNMAFSPF SIASLLTQVL LGAGENTKTN LESILSYPKD FTCVHQALKG FTTKGVTSVS QIFHSPDLAI RDTFVNASRT LYSSSPRVLS NNSDANLELI NTWVAKNTNN KISRLLDSLP SDTRLVLLNA IYLSAKWKTT FDPKKTRMEP FHFKNSVIKV PMMNSKKYPV AHFIDQTLKA KVGQLQLSHN LSLVILVPQN LKHRLEDMEQ ALSPSVFKAI MEKLEMSKFQ PTLLTLPRIK VTTSQDMLSI MEKLEFFDFS YDLNLCGLTE DPDLQVSAMQ HQTVLELTET GVEAAAASAI SVARTLLVFE VQQPFLFVLW DQQHKFPVFM GRVYDPRA
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less