missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BRWD2 (aa 268-348) Control Fragment Recombinant Protein

Catalog No. RP97074
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97074 Supplier Invitrogen™ Supplier No. RP97074

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58620 (PA5-58620. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

WDR11 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is located in the chromosome 10q25-26 region, which is frequently deleted in gliomas and tumors of other tissues, and is disrupted by the t(10;19) translocation rearrangement in glioblastoma cells. The gene location suggests that it is a candidate gene for the tumor suppressor locus.

Specifications

Accession Number Q9BZH6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55717
Name Human BRWD2 (aa 268-348) Control Fragment
Quantity 100 μL
Gene Alias 2900055P10Rik; AW489876; bromodomain and WD repeat domain containing 2; bromodomain and WD repeat-containing protein 2; BRWD2; DR11; HH14; Kiaa1351; mKIAA1351; sensitization to ricin complex subunit 1; SRI1; WD repeat domain 11; WD repeat domain 15; WD repeat-containing protein 11; WD repeat-containing protein 15; WDR11; WDR15
Common Name BRWD2
Gene Symbol WDR11
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ILILDLEVNQTVGVIAIERTGVPFLQVIPCFQRDGLFCLHENGCITLRVRRSYNNIFTTSNEEPDPDPVQELTYDLRSQCD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less