missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BRUNOL4 (aa 314-369) Control Fragment Recombinant Protein

Catalog No. RP108723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108723 Supplier Invitrogen™ Supplier No. RP108723
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139747 (PA5-139747. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Accession Number Q9BZC1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56853
Name Human BRUNOL4 (aa 314-369) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A230070D14Rik; Brul4; Bruno -like 4; brunol4; BRUNOL-4; bruno-like 4, RNA binding protein; bruno-like protein 4; C130060B05Rik; celf4; CELF-4; CUGBP; CUG-BP and ETR-3 like factor 4; CUG-BP- and ETR-3-like factor 4; CUGBP Elav-like family member 4; CUGBP, Elav-like family member 4; LYST-interacting protein LIP9; RNA-binding protein BRUNOL4; RNA-binding protein BRUNOL-4; zgc:92761
Common Name BRUNOL4
Gene Symbol CELF4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PTSGGSTPPGITAPAVPSIPSPIGVNGFTGLPPQANGQPAAEAVFANGIHPYPAQS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less