missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-7 Recombinant Protein DFS Item

Recombinant Protein

$416.00 - $3,310.00

Specifications

Accession Number P18075, P23359
For Use With (Application) Bioactivity
Formulation 10mM acetic acid with no preservative
Gene ID (Entrez) 12162, 655
Molecular Weight (g/mol) 38 kDa
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC9541 DFS Item
View Documents
Gibco™
PHC9541
100 μg N/A
Only null left
 
PHC9543 DFS Item
View Documents
Gibco™
PHC9543
1 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PHC9544 DFS Item
View Documents
Gibco™
PHC9544
10 μg N/A
Only null left
 
Description

Description

Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile water containing 0.1% human serum albumin. Apportion the reconstituted protein into working aliquots and store at = -20°C. Make any further dilutions of the reconstituted protein in low endotoxin medium or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA. Storage: Store the lyophilized protein at = -20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at = -20°C. Avoid repeated freeze-thaw cycles.

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
Specifications

Specifications

P18075, P23359
10mM acetic acid with no preservative
38 kDa
Purified
RUO
BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5
Bmp7
Protein
Human
-20°C
ED50 = 8.31 ng/mL; determined by the dose-dependent induction of alkaline phosphatase production in ATDC-5 cells.
<0.5 EU/μg
HEK293 cells
Recombinant, Ligand
BMPs (Bone Morphogenetic Proteins) & Related Factors
Gibco
Bioactivity
12162, 655
Human BMP-7
-20°C
<0.5 EU/μg
BMP-7
ED50 = 8.31 ng/mL; determined by the dose-dependent induction of alkaline phosphatase production in ATDC-5 cells.
Unconjugated
Recombinant
Carrier-Free
Induction of alkaline phosphatase in ATDC5 cells
Dry Ice
Growth Factors & Receptors
Lyophilized
Bone Research, Stem Cell Research, Differentiation
>95% by SDS-PAGE
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'