Learn More
Gibco™ Human BMP-4, Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
$338.00 - $22633.60
Specifications
| Accession Number | P12644 |
|---|---|
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 652 |
| Name | Human BMP-4, Animal-Free |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AF12005ET
|
Gibco™
AF12005ET500UG |
500 μg |
Each for $3,900.00
|
|
|||||
|
50-272-2975
|
Gibco™
AF12005ET01M |
2 x 500 μg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AF12005ET10
|
Gibco™
AF12005ET10UG |
10 μg |
Each for $338.00
|
|
|||||
Description
AF-120-05ET-1MG will be provided as 2 x 500 μg (AF-120-05ET-500UG). PeproTech's E.coli-derived BMP-4 is a fully active homodimeric protein consisting of two 107 amino acid subunits, which correspond to amino acids 303-408 of the full length BMP-4 precursor, and includes a N-terminal Met residue. The calculated molecular weight of Recombinant Human BMP-4 (E.coli-derived) is 24.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.Specifications
| P12644 | |
| Protein with no preservative | |
| Human BMP-4, Animal-Free | |
| RUO | |
| BMP4 | |
| Recombinant | |
| Lyophilized |
| Control, Bioactivity | |
| 652 | |
| E. coli | |
| BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME | |
| Unconjugated | |
| MKKNKNCRRH SLYVDFSDVG WNDWIVAPPG YQAFYCHGDC PFPLADHLNS TNHAIVQTLV NSVNSSIPKA CCVPTELSAI SMLYLDEYDK VVLKNYQEMV VEGCGCR | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |