missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human beta-Thromboglobulin Native Protein
Description
RP-43150 contains native human b-Thromboglobulin protein. RP-43150 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.
Specifications
Specifications
| Accession Number | P02775 |
| For Use With (Application) | Neutralization |
| Formulation | 0.025M HEPES with 0.15M NaCl and no preservative; pH 7.4 |
| Gene ID (Entrez) | 5473 |
| Molecular Weight (g/mol) | 35.8 kDa |
| Name | Human beta-Thromboglobulin |
| pH Range | 7.4 |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Source | Human |
| Show More |