missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human beta-Glucuronidase/GUSB ELISA Kit

Catalog No. EH43RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH43RBX5 5 x 96 Tests
EH43RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH43RBX5 Supplier Invitrogen™ Supplier No. EH43RBX5
Only null left

ELISA

Human beta-Glucuronidase/GUSB quantitates human beta-Glucuronidase/GUSB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human beta-Glucuronidase/GUSB.

The GUSB gene encodes beta-glucuronidase (EC 3.2.1.31), a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans. It is a tetrameric glycoprotein composed of identical subunits. The GUSB gene is mutated in mucopolysaccharidosis type VII (MPS7; MIM 253220).

Specifications

Accession Number Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; GUSB; Gus-r; Gus-s; Gus-t; Gus-u; Gut; MPS7
Assay Range 0.18 to 40 ng/mL
Assay Sensitivity 0.18 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2990
Gene Symbol GUSB
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GUSB
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less