Learn More
Invitrogen™ Human beta-2 Defensin ELISA Development Kit (TMB), PeproTech®
ELISA
Supplier: Invitrogen™ 900T172K
Description
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.Specifications
| O15263 | |
| Lot-specific | |
| ELISA | |
| 1673 | |
| Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
| 10 x 96 Tests | |
| Refer to product documentation for component specific storage temperature. | |
| 1 hrs 45 mins |
| Lot-specific | |
| HRP | |
| Colorimetric Microplate Reader | |
| DEFB4A | |
| HRP | |
| RUO | |
| beta-2 Defensin | |
| 24 hrs 30 mins |