missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BAIAP2L2 (aa 351-499) Control Fragment Recombinant Protein

Catalog No. RP91512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91512 Supplier Invitrogen™ Supplier No. RP91512

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51807. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene binds phosphoinositides and promotes the formation of planar or curved membrane structures. The encoded protein is found in RAB13-positive vesicles and at intercellular contacts with the plasma membrane. [provided by RefSeq, Dec 2012].

Specifications

Accession Number Q6UXY1
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 80115
Name Human BAIAP2L2 (aa 351-499) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BAI1 associated protein 2 like 2; BAI1-associated protein 2-like 2; BAI1-associated protein 2-like protein 2; Baiap2l2; BC048937; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 2; pinkbar; Planar intestinal- and kidney-specific BAR domain protein; RGD1308786; UNQ9336/PRO34007
Common Name BAIAP2L2
Gene Symbol BAIAP2L2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VVEVLVPEAQNGWLYGKLEGSSASGWFPEAYVKALEEGPVNPMTPVTPMTSMTSMSPMTPMNPGNELPSRSYPLRGSHSLDDLLDRPGNSIAPSEYWDGQSRSRTPSRVPSRAPSPAPPPLPSSRRSSMGSTAVATDVKKLMSSEQYPP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less