missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ASF1A (aa 175-201) Control Fragment Recombinant Protein

Catalog No. RP107819
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107819 Supplier Invitrogen™ Supplier No. RP107819

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair.

Specifications

Accession Number Q9Y294
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25842
Name Human ASF1A (aa 175-201) Control Fragment
Quantity 100 μL
Gene Alias 2310079C17Rik; anti-silencing function 1 A histone chaperone; anti-silencing function protein 1 homolog A; ASF1 anti-silencing function 1 homolog A; ASF1A; C79052; CCG1-interacting factor A; CGI-98; CIA; DKFZP547E2110; hAsf1; hAsf1a; hCIA; histone chaperone ASF1A; HSPC146; RP3-329L24.1
Common Name ASF1A
Gene Symbol ASF1A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence STDALPSASKGWSTSENSLNVMLESHM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less