missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ARHGEF37 (aa 181-250) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60542 (PA5-60542. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May act as a guanine nucleotide exchange factor (GEF).
Specifications
Specifications
| Accession Number | A1IGU5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 389337 |
| Name | Human ARHGEF37 (aa 181-250) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4933429F08Rik; ARHGEF37; probable guanine nucleotide exchange factor FLJ41603 homolog; protein 2-88; RGD1560471; Rho guanine nucleotide exchange factor (GEF) 37; Rho guanine nucleotide exchange factor 37; Scaffold protein tuba 3 |
| Common Name | ARHGEF37 |
| Show More |