missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ARHGAP19 (aa 280-376) Control Fragment Recombinant Protein

Catalog No. RP105908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105908 Supplier Invitrogen™ Supplier No. RP105908
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65159 (PA5-65159. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Members of the ARHGAP family, such as ARHGAP19, encode negative regulators of Rho GTPases (see RHOA; MIM 165390), which are involved in cell migration, proliferation, and differentiation, actin remodeling, and G1 cell cycle progression.

Specifications

Accession Number Q14CB8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84986
Name Human ARHGAP19 (aa 280-376) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4933411B03Rik; AI120128; ARHGAP19; AW546831; mFLJ00368; putative RhoGAP protein; Rho GTPase activating protein 19; rho GTPase-activating protein 19; Rho-type GTPase-activating protein 19
Common Name ARHGAP19
Gene Symbol ARHGAP19
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NVTANDLQENITKLNSGMAFMIKHSQKLFKAPAYIRECARLHYLGSRTQASKDDLDLIASCHTKSFQLAKSQKRNRVDSCPHQEETQHHTEEALREL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less