missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human APOLD1 (aa 139-192) Control Fragment Recombinant Protein

Catalog No. RP97864
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97864 Supplier Invitrogen™ Supplier No. RP97864
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62616 (PA5-62616. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in angiogenesis. May play a role in activity-dependent changes of brain vasculature. May affect blood-brain permeability.

Specifications

Accession Number Q96LR9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 81575
Name Human APOLD1 (aa 139-192) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias APOLD1; apolipoprotein L domain containing 1; apolipoprotein L domain-containing protein 1; Vascular early response gene protein; VERGE
Common Name APOLD1
Gene Symbol APOLD1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SLIFCNSRELRRVQEIAATCQDQMREILSCLEFFCRWQGCGDRQLLQCGRNASI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less