missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ANR52 (aa 494-540) Control Fragment Recombinant Protein

Catalog No. RP100403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100403 Supplier Invitrogen™ Supplier No. RP100403
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60523 (PA5-60523. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. [UniProt]

Specifications

Accession Number Q8NB46
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 283373
Name Human ANR52 (aa 494-540) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6430544C07; ANKRD33; ANKRD52; ankyrin repeat domain 33; ankyrin repeat domain 52; ankyrin repeat domain-containing protein 52; CVWG5837; G431002C21Rik; PP6-ARS-C; protein phosphatase 6 ankyrin repeat subunit C; RGD1307124; Serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C; serine/threonine-protein phosphatase 6 regulatory subunit ARS-C
Common Name ANR52
Gene Symbol ANKRD52
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HYAAASDTYRRAEPHTPSSHDAEEDEPLKESRRKEAFFCLEFLLDNG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less