missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; Anx1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; Calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; phospholipase A2 inhibitory protein; RP11-71A24.1 |
|---|---|
| Assay Range | 1.64 to 400 ng/mL |
| Assay Sensitivity | 1.6 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH31RBX5
|
Invitrogen™
EH31RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH31RB
|
Invitrogen™
EH31RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Annexin A1 quantitates human Annexin A1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Annexin A1.
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.Specifications
| Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; Anx1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; Calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; phospholipase A2 inhibitory protein; RP11-71A24.1 | |
| 1.6 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| ANXA1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Annexin A1 | |
| 4 hrs 45 mins |
| 1.64 to 400 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 301 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |