missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AmiGO2 ELISA Kit
ELISA
Supplier: Invitrogen™ EH23RBX5
Description
Human AmiGO2 quantitates human AmiGO2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human AmiGO2.
A novel primary response gene, Alivin 1 (ali1), is an activity-dependent gene and promotes survival of neurons. Sequence analyses reveal that rat, mouse, and human Ali1 proteins contain seven leucine-rich repeats, one IgC2-like loop and a transmembrane domain, and display homology to Kek and Trk families. Results suggest that expression of ali1 promotes depolarization-dependent survival of the granule neuron. Mouse ali1 was mapped to a locus approximately 55.3 cM from the centromere on chromosome 15 that is syntenic to positional candidate loci for familial Alzheimer's disease type 5 and Parkinson's disease 8 on human chromosome 12.Chemical Identifiers
| adhesion molecule with Ig like domain 2; adhesion molecule with Ig-like domain 2; AI415330; ALI1; alivin 1; alivin-1; AMIGO2; AMIGO-2; amigo2 {ECO:0000312; amphoterin induced gene 2; amphoterin induced gene and ORF 2; amphoterin-induced protein 2; AW208913; DEGA; differentially expressed in gastric adenocarcinoma; differentially expressed in gastric adenocarcinomas; EMBL:AAO48951.1}; transmembrane protein AMIGO2 | |
| 0.2 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Amigo2 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Alivin 1 | |
| 4 hrs 45 mins |
| 0.2 to 50 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 347902 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| adhesion molecule with Ig like domain 2; adhesion molecule with Ig-like domain 2; AI415330; ALI1; alivin 1; alivin-1; AMIGO2; AMIGO-2; amigo2 {ECO:0000312; amphoterin induced gene 2; amphoterin induced gene and ORF 2; amphoterin-induced protein 2; AW208913; DEGA; differentially expressed in gastric adenocarcinoma; differentially expressed in gastric adenocarcinomas; EMBL:AAO48951.1}; transmembrane protein AMIGO2 | |
| 0.2 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Amigo2 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.2 to 50 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 347902 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 5 x 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Alivin 1 | |
| 4 hrs 45 mins |