missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Alpha-Galactosidase A/GLA ELISA Kit
ELISA
Specifications
| Accession Number | Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; melibiase |
|---|---|
| Assay Range | 45 to 10,000 pg/mL |
| Assay Sensitivity | 45 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Description
Human Alpha-Galactosidase A/GLA quantitates human Alpha-Galactosidase A/GLA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Alpha-Galactosidase A/GLA.
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predominantly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.Specifications
| Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; melibiase | |
| 45 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| GLA | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| alpha Galactosidase | |
| 4 hrs 45 mins |
| 45 to 10,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 2717 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |