missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ALK-6/BMPR-IB ELISA Kit

Catalog No. EH18RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH18RBX5 5 x 96 Tests
EH18RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH18RBX5 Supplier Invitrogen™ Supplier No. EH18RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions

ELISA

Human ALK-6/BMPR-IB quantitates human ALK-6/BMPR-IB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ALK-6/BMPR-IB.

BMPR1B (also known as ALK6, CDw293) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.

Specifications

Accession Number O00238
Assay Range 24-6000 pg/mL
Assay Sensitivity 24 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Activin receptor-like kinase 6; Acvrlk6; AI385617; Alk6; ALK-6; AMDD; AV355320; BDA1D; BDA2; BMP type-1B receptor; BMP15; Bmpr1b; BMPR-1B; BMPRIB; BMPR-IB; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; bone morphogenetic protein receptor type 1B; bone morphogenetic protein receptor type IB; bone morphogenetic protein receptor type-1B; bone morphogenetic protein receptor, type 1B; bone morphogenetic protein receptor, type IB; CDw293; CFK-43a; serine/threonine receptor kinase; serine/threonine-protein kinase receptor R6; SKR6Activin receptor-like kinase 6; Acvrlk6; AI385617; Alk6; ALK-6; AMDD; AV355320; BDA1D; BDA2; BMP type-1B receptor; BMP15; Bmpr1b; BMPR-1B; BMPRIB; BMPR-IB; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; bone morphogenetic protein receptor type 1B; bone morphogenetic protein receptor type IB; bone morphogenetic protein receptor type-1B; bone morphogenetic protein receptor, type 1B; bone morphogenetic protein receptor, type IB; CDw293; CFK-43a; serine/threonine receptor kinase; serine/threonine-protein kinase receptor R6; SKR6;
Gene ID (Entrez) 658
Gene Symbol BMPR1B
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CDw293
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less