missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human ALDH1A1 (NP_000680.2) VersaClone cDNA

Specifications
Specifications
| Gene | Aldehyde Dehydrogenase 1-A1/ALDH1A1 |
| Accession Number | NP_000680.2 |
| Gene ID (Entrez) | 216 |
| Gene Alias | acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde Dehydrogenase 1A1, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH1A1, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |