missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AKNA (aa 118-221) Control Fragment Recombinant Protein

Catalog No. RP106388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106388 Supplier Invitrogen™ Supplier No. RP106388
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (53%), Rat (53%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65717 (PA5-65717. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

AKNA is a transcription factor that specifically activates the expression of the CD40 receptor and its ligand CD40L/CD154, two cell surface molecules on lymphocytes that are critical for antigen-dependent-B-cell development. It binds to A/T-rich promoters.

Specifications

Accession Number Q7Z591
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80709
Name Human AKNA (aa 118-221) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AKNA; AKNA transcript F2; AT-hook transcription factor; AT-hook transcription factor AKNA; AT-hook-containing transcription factor; KIAA1968; Microtubule organization protein AKNA
Common Name AKNA
Gene Symbol AKNA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LDMTEEEPDGTLGSLEVEEAGESSSRLGYEAGLSLEGHGNTSPMALGHGQARGWVASGEQASGDKLSEHSEVNPSVELSPARSWSSGTVSLDHPSDSLDSTWEG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less