missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Adiponectin ProQuantum Immunoassay Kit
ELISA
$920.00 - $4140.00
Specifications
| Accession Number | Q15848 |
|---|---|
| Assay Range | 0.76 to 1,500,000 pg/mL |
| Assay Sensitivity | <0.7 pg/mL |
| Conjugate | FAM™ |
| Detection Method | Taqman™ Real-time PCR |
Description
Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.Specifications
| Q15848 | |
| <0.7 pg/mL | |
| Taqman™ Real-time PCR | |
| ProQuantum | |
| Human | |
| qPCR Instruments | |
| 1 hr. | |
| FAM™ | |
| Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
| Adiponectin | |
| 2 hrs |
| 0.76 to 1,500,000 pg/mL | |
| FAM™ | |
| Proximity-based Immunoassay | |
| Plasma, Serum, Supernatant | |
| Protein Quantitation | |
| 9370 | |
| Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
| RUO | |
| Frozen at -20°C | |
| 30 mins |