missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Adiponectin ProQuantum Immunoassay Kit

Description
Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.
Specifications
Specifications
| Accession Number | Q15848 |
| Assay Range | 0.76 to 1,500,000 pg/mL |
| Assay Sensitivity | <0.7 pg/mL |
| Conjugate | FAM™ |
| Detection Method | Taqman™ Real-time PCR |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | Protein Quantitation |
| Show More |