missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ADAMTS4 ELISA Kit
Description
Human ADAMTS4 quantitates human ADAMTS4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ADAMTS4.
This gene encodes a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene lacks a C-terminal TS motif. It is responsible for the degradation of aggrecan, a major proteoglycan of cartilage, and brevican, a brain-specific extracellular matrix protein. The cleavage of aggrecan and brevican suggests key roles of this enzyme in arthritic disease and in the central nervous system, potentially, in the progression of glioma.
Specifications
Specifications
| Accession Number | 3830423K05; a disintegrin and metallopeptidase with thrombospondin motifs 4; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 4; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 4; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 4; ADAM metallopeptidase with thrombospondin type 1 motif, 4; ADAMs; ADAM-TS 4; ADAMTS family member; ADAMTS-2; Adamts4; ADAM-TS4; ADAMTS-4; ADMP-1; Aggrecanase; Aggrecanase 1; aggrecanase-1; KIAA0688; metalloendopeptidases; metalloproteinase; mKIAA0688; UNQ769/PRO1563 |
| Assay Range | 0.12 to 30 ng/mL |
| Assay Sensitivity | 0.12 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 9507 |
| Gene Symbol | ADAMTS4 |
| Interassay CV | <12% |
| Show More |