missing translation for 'onlineSavingsMsg'
Learn More

Human ADAM10 Ectodomain APC-conjugated Antibody, R&D Systems™

Catalog No. IC1427A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1427A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC1427A Supplier Bio-Techne Supplier No. IC1427A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for Flow Cytometry∣Flow Cytometry (Intracellular Staining)∣Immunohistochemistry (Frozen).

Specifications

Antigen ADAM10
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 163003
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O14672
Gene Alias a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian, Kuzbanian protein homolog, MADM, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease
Gene Symbols ADAM10
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ADAM10 Thr214-Glu672 Accession # O14672
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102
Test Specificity Detects human ADAM10 in direct ELISAs and Western blots. In Western blots, shows less than 5% cross-reactivity with recombinant human (rh) ADAM8, 9, 15, 17/TACE, and recombinant mouse ADAM10.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line R&D Systems
Show More Show Less