missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ACY3 Control Fragment Recombinant Protein

Catalog No. RP104141
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104141 Supplier Invitrogen™ Supplier No. RP104141

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61683. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.

Specifications

Accession Number Q96HD9
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 91703
Name Human ACY3 Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 0610006H10Rik; AA3; AAIII; Acy3; ACY-3; Acylase III; aminoacylase 3; Aminoacylase III; Aminoacylase-3; Aspa2; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; aspartoacylase-2; aspartoacylase-3; AW107362; EGK_05921; HCBP1; HCV core-binding protein 1; hepatitis C virus core-binding protein 1; LOW QUALITY PROTEIN: N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); RGD1305394
Common Name ACY3
Gene Symbol Acy3
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MCSLPVPQEPLRRVAVTGGTHGNEMSGVYLARHWLHAPAELQRASFSAVPVLANPAATSGCRRYVDHDLNRTFTSSFLNSR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less