missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Activin B (NP_002184.2) VersaClone cDNA

Specifications
Specifications
| Accession Number | NP_002184.2 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |