missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ACSBG2 (aa 559-635) Control Fragment Recombinant Protein

Catalog No. RP99078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99078 Supplier Invitrogen™ Supplier No. RP99078

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60359 (PA5-60359. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Catalyzes the conversion of fatty acids such as long chain and very long-chain fatty acids to their active form acyl-CoAs for both synthesis of cellular lipids, and degradation via beta-oxidation. Can activate diverse saturated, monosaturated and polyunsaturated fatty acids (PubMed:16371355, PubMed:16762313). Has increased ability to activate oleic and linoleic acid (PubMed:16371355). May play a role in spermatogenesis (PubMed:15685348). [UniProt]

Specifications

Accession Number Q5FVE4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 81616
Name Human ACSBG2 (aa 559-635) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ACSBG2; Acyl-CoA synthetase bubblegum family member 2; Arachidonate--CoA ligase ACSBG2; BGR; BRGL; bubblegum-related protein; Long-chain-fatty-acid--CoA ligase ACSBG2; PRTDNY3; PRTD-NY3; testicular tissue protein Li 8; UNQ2443/PRO5005
Common Name ACSBG2
Gene Symbol ACSBG2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LKCEMNQMSGEPLDKLNFEAINFCRGLGSQASTVTEIVKQQDPLVYKAIQQGINAVNQEAMNNAQRIEKWVILEKDF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less