Learn More
Velp™ HU6 Hydrolysis Unit
Description
- Solution for sample preparation frees fat molecules so they are ready for extraction
- Procedure is required in a number of AOAC Official Methods to determine the fat content of food samples such as meat, cheese, seafood, chocolate, cereal flours, etc.
- Performs hydrolysis in safe fashion and handles six samples at a time to maximize productivity
- Supplied with vacuum pump to reduce acid fumes
- Provides effective filtration and rinsing of 3 to 6 samples in parallel
- Valves allow user to selectively interrupt the vacuum in each position, speeding up the filtration process
Specifications
Specifications
| Product Type | Hydrolysis Unit |
| For Use With (Equipment) | HU6 Solvent Extractor |
| Frequency | 50/60 Hz |
| Voltage | 115 V |