Learn More
Falcon™ HTS 24-well Multiwell Permeable Support System with High Density PET Membrane
Description
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery.
- Available in a choice of membrane pore sizes, these multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays.
- These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers.
- Single unit that is compatible with all Falcon 24 well plates and feeder trays.
Specifications
Specifications
| Sterility | Sterile |
| Type | Cell Culture Insert |
| For Use With (Application) | Bioavailability Studies, Transport Studies, Permeability Studies, Cell Migration Studies, and Tumor Invasion Studies |
| No. per Case | 5 Inserts |
| No. per Pack | 5 Inserts |
| Pore Size | 3.0 μm |