missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA585605
Description
Keep as concentrated solution. Positive Control: SH-SY5Y, SH-SY5Y membrane fraction extract, unboiled SH-SY5Y, unboiled SH-SY5Y membrane fraction extract, mouse liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HTR2B is a g-protein coupled receptor for 5-hydroxytryptamine (serotonin). It functions as a receptor for various ergot alkaloid derivatives and psychoactive substances. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors. HTR2B also plays a role in perception of pain, regulation of behavior. It is required to normal proliferation of embryonic cardiac myocytes and normal heart development.Specifications
| HTR2B | |
| Polyclonal | |
| Unconjugated | |
| HTR2B | |
| 5-HT 2B receptor, 5-HT(2B), 5-HT2B, 5-HT-2B, 5HT2B Receptor, 5-HT2b receptor, 5-HT-2F, 5-hydroxytryptamine (serotonin) receptor 2B, 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled, 5-hydroxytryptamine 2B receptor, 5-hydroxytryptamine receptor 2B, 5-hydroxytryptamine receptor 2B variant b, AJ012488, AV377389, HTR2B, NP75 protein, serotonin receptor 2B, Srl, Stomach fundus serotonin receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15559, 3357 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2.29 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300, pH 7 | |
| P41595, Q02152 | |
| HTR2B | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human 5-HT2B receptor. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |