missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HTR1B Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA120650

Catalog No. PA120650

Only null left
Explore available promotions

Description

Description

PA1-20650 detects 5-HT1B Receptor in human samples.

Serotonin (5-hydroxytryptamine, 5-HT), originally discovered as a serum factor plays important roles in regulating diverse biological processes in central and peripheral nervous systems, cardiovascular systems, and gastrointestinal systems. Molecular cloning studies have identified 5 receptors belonging to the 5-HT1 subtypes. These receptors are negatively coupled to adenylyl cyclase. Radioligand binding studies suggest the presence of 5-HT1B (formerly known as 5-HT1DbR) sites in substantia nigra, dorsal subiculum, globus pallidus, and superior collicum. In situ hybridization studies show abundance of 5-HT1BR mRNA in CA1 pyramidal cells of hippocampus, Purkinje cells of cerebellum, olfactory tubercle, subthalamic nucleus and somewhat lower levels in the caudate-putamen, cortical areas, thalamic and hypothalamic nuclei.
Specifications

Specifications

HTR1B
Polyclonal
Unconjugated
HTR1B
5-HT>1B> receptor; 5ht1b; 5-HT1B; 5-HT-1B; 5HT1B Receptor; 5-HT1B receptor; 5-HT1B serotonin receptor; 5-HT1DB; 5-HT-1D-beta; 5-hydroxytryptamine (serotonin ) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled; 5-hydroxytryptamine receptor 1B; HTR1B; HTR1D2; HTR1DB; S12; serotonin (5-hydroxytryptamine (5HT)) receptor type 1B; serotonin 1D beta receptor; Serotonin receptor 1B
Rabbit
Antigen affinity chromatography
RUO
100009594, 100689321, 100717602, 15551, 25075, 317707, 3351, 397563, 403741
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
O08892, P28222, P28334, P28564, P46636, P49144, P79250, P79399
HTR1B
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human 5HT1B Receptor.
25 μg
Primary
Human, Mouse, Rat, Canine, Rabbit, Bovine, Hamster, Pig, Guinea Pig, Bat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions