missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ HT 96-well Electroporation Plate

Description
- 96-well disposable electroporation plate
- 2 mm gap
- 100 μL
- Coated
- 1 plate
Specifications
Specifications
| Format | 96-well |
| For Use With (Application) | Transfection of Mammalian Cell, Transformation of Bacteria and Yeast, siRNA and cDNA Library Screening |
| For Use With (Equipment) | ECM 630 HT, ECM 830 HT, Gemini X2 HT, 96-Well Electroporation Systems |
| Product Line | BTX Electroporation |
| Product Type | Electroporation Consumable |
| Description | 96-well Disposable Electroporation Plate, 2 mm gap, 100 μl, Coated, 1 plate |
| Electroporation Chamber | 96-well Electroporation Plate |
| Quantity | 1 Ea. |
| Size (English) Gap | 0.078 in. |
| Size (Metric) Gap | 2 mm |
| Show More |