missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSPA1A Monoclonal Antibody (K40046_8C11), Invitrogen™

Catalog No. MA555917
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555917 50 μL
MA556297 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555917 Supplier Invitrogen™ Supplier No. MA555917
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: QDFFNGRDLN KSINPDEAVA YGAAVQAAIL MGDKSENVQD LLLLDVAPLS LGLETAGGVM TALIKRNSTI PTKQTQIFTT YSDNQPGVLI QVYEGERAMT KDNNLLGRFE LSGIPPAPRG VPQIEVTFDI DANGILNVTA TDKSTGKANK ITITNDKGRL SKEEIERMVQ EAEKYKAEDE VQRERVSAKN ALESYAFNMK SAVEDEGLKG KISEADKKKV LDKCQEVISW LDANTLAEKD EFEHKRKELE

This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.

Specifications

Antigen HSPA1A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K40046_8C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene HSPA1A
Gene Accession No. P0DMV8
Gene Alias 68 kDa heat shock protein; APG2; APG-2; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70 kDa protein 4; heat shock 70kD protein 1A; heat shock 70kD protein 4; heat shock 70kDa protein 1A; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; heat shock protein 1A; heat shock protein 70; heat shock protein 70-1; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 1A; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; heat shock protein, 70 kDa 3; heat shock-induced protein; HEL-S-103; HEL-S-5a; HS24/P52; hsp68; hsp70; hsp70 RY; HSP70.1; HSP70.1/2; HSP70.1/HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; Hsp70-2; Hsp70-3; Hsp70A1; HSP70I; hsp70RY; HSP72; Hspa1; HSPA1A; Hspa1b; Hspa2; HSPA4; HSPH2; HSX70; inducible heat shock protein 70; RY
Gene Symbols HSPA1A
Host Species Mouse
Immunogen Recombinant human HSPA1A (immunogen range is 351-600).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3303
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less