missing translation for 'onlineSavingsMsg'
Learn More

5 Percent Platinum on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical

Catalog No. 18607962
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18607962 100 g
18607960 5 g
18607961 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607962 Supplier Spectrum Chemical Mfg Cor Supplier No. SP204100G
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
IUPAC Name platinum
SMILES [Pt]

Specifications

Physical Form Powder or Liquid
Quantity 100 g
Grade Not Available
Packaging Amber Glass Bottle
Chemical Name or Material 5% Platinum on Alumina Powder, Reduced, Dry, 70 μ, BASF Catalyst

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.