missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSD17B6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579406
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579406 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579406 Supplier Invitrogen™ Supplier No. PA579406
Only null left

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Human Placenta Tissue, MCF-7 whole cell. IHC: human liver cancer tissue.

The protein encoded by this gene has both oxidoreductase and epimerase activities and is involved in androgen catabolism. The oxidoreductase activity can convert 3 alpha-adiol to dihydrotestosterone, while the epimerase activity can convert androsterone to epi-androsterone. Both reactions use NAD+ as the preferred cofactor. This gene is a member of the retinol dehydrogenase family. Transcript variants utilizing alternative polyadenylation signals exist.

Specifications

Antigen HSD17B6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene HSD17B6
Gene Accession No. O14756
Gene Alias 17-beta-HSD 6; 17-beta-HSD6; 17betaHSD9; 17-beta-HSD9; 17-beta-hydroxysteroid dehydrogenase type 6; 3(alpha->beta)-hydroxysteroid epimerase; 3(alpha->beta)-hydroxysteroid epimerasel; 3-alpha->beta-HSE; 3-alpha->beta-hydroxysteroid epimerase; 3-hydroxysteroid epimerase; Gm182; Hsd17b6; Hsd17b9; HSE; hydroxysteroid (17-beta) dehydrogenase 6; hydroxysteroid (17-beta) dehydrogenase 6 homolog; hydroxysteroid (17-beta) dehydrogenase 9; hydroxysteroid 17-beta dehydrogenase 6; hydroxysteroid 17-beta dehydrogenase 9; hypothetical protein LOC533086; NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase; oxidative 17 beta hydroxysteroid dehydrogenase type 6; oxidative 3-alpha hydroxysteroid dehydrogenase; oxidative 3-alpha-hydroxysteroid-dehydrogenase; oxidoreductase; Rdh8; retinol dehydrogenase; RODH; SDR9C6; Short chain dehydrogenase/reductase family 9C member 6; short chain dehydrogenase/reductase family 9C, member 6
Gene Symbols HSD17B6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human HSD17B6 (300-317aa SLADYILTRSWPKPAQAV).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8630
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less